6-Amino-3-azabicyclo[3.1.1]heptane-3-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

115743-250MG 115743-500MG 115743-1G
102897-312EA 2143.08 USD
102897-312 102897-314 102897-346
6-Amino-3-azabicyclo[3.1.1]heptane-3-carboxylic acid tert-butyl ester ≥95%
6-Amino-3-azabicyclo[3.1.1]heptane-3-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD17168422
CAS Number: 1250997-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR