1-Cyclopropyl-4-iodo-1H-pyrazole ≥95%

Supplier: Matrix Scientific

115742-500MG 115742-1G 115742-5G
102899-646EA 413.85 USD
102899-646 102899-648 102899-650
1-Cyclopropyl-4-iodo-1H-pyrazole ≥95%
1-Cyclopropyl-4-iodo-1H-pyrazole

Hazard: Irritant

Formula: C₆H₇IN₂
MDL Number: MFCD22573364
CAS Number: 1239363-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR