1-Azetidinesulfonyl chloride 94

Supplier: Matrix Scientific

Synonyms: Azetidine-1-sulphonyl chloride

115741-100MG 115741-500MG 115741-1G
102897-306EA 330.77 USD
102897-306 102897-308 102897-310
1-Azetidinesulfonyl chloride 94
1-Azetidinesulfonyl chloride

Hazard: Irritant

Formula: C₃H₆ClNO₂S
MW: 155.6 g/mol
MDL Number: MFCD19200214
CAS Number: 639519-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR