tert-Butyl-3,3-difluoro-4-(hydroxymethyl)-4-methylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115740-250MG 115740-1G
102897-302EA 1867.69 USD
102897-302 102897-304
tert-Butyl-3,3-difluoro-4-(hydroxymethyl)-4-methylpiperidine-1-carboxylate ≥95%
tert-Butyl-3,3-difluoro-4-(hydroxymethyl)-4-methylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁F₂NO₃
MW: 265.3 g/mol
CAS Number: 1334412-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR