7-Thieno[3,2-d]pyrimidine ≥97%

Supplier: Matrix Scientific

Synonyms: Thieno[3,2-d]pyrimidine

115739-1G 115739-5G 115739-25G
102897-296EA 330.77 USD
102897-296 102897-298 102897-300
7-Thieno[3,2-d]pyrimidine ≥97%
7-Thieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆H₄N₂S
MW: 136.18 g/mol
MDL Number: MFCD11110439
CAS Number: 272-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR