Bis(2,4,6-trimethylpyridine)silver(I) hexafluorophosphate ≥95%

Supplier: Matrix Scientific

115735-1G 115735-5G
102897-274EA 140 USD
102897-274 102897-276
Bis(2,4,6-trimethylpyridine)silver(I) hexafluorophosphate ≥95%
Bis(2,4,6-trimethylpyridine)silver(I) hexafluorophosphate

Hazard: Irritant

Formula: C₁₀H₁₉AgF₆NP
CAS Number: 271590-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR