tert-Butyl-2-(tetrahydro-4H-pyran-4-ylidene)acetate ≥95%

Supplier: Matrix Scientific

115731-5G 115731-1G
102897-264EA 2792.31 USD
102897-264 102900-232
tert-Butyl-2-(tetrahydro-4H-pyran-4-ylidene)acetate ≥95%
tert-Butyl-2-(tetrahydro-4H-pyran-4-ylidene)acetate

Hazard: Irritant

Formula: C₁₁H₁₈O₃
MDL Number: MFCD22415212
CAS Number: 894789-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR