Methyl-1-methyl-4-nitro-1H-pyrazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 1-methyl-4-nitro-1H-pyrazole-5-carboxylate
115725-1G
115725-5G
102899-492EA
798.46
USD
102899-492
102899-608
Methyl-1-methyl-4-nitro-1H-pyrazole-5-carboxylate ≥95%
Methyl-1-methyl-4-nitro-1H-pyrazole-5-carboxylate
Hazard: Irritant
Formula:
C₆H₇N₃O₄ MW: 185.14 g/mol |
MDL Number:
MFCD02110822 CAS Number: 309740-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...