3-Fluoro-1-[tris(1-methylethyl)silyl]-1H-pyrrole ≥95%

Supplier: Matrix Scientific

115712-1G 115712-5G
102899-598EA 989.23 USD
102899-598 102899-600
3-Fluoro-1-[tris(1-methylethyl)silyl]-1H-pyrrole ≥95%
3-Fluoro-1-[tris(1-methylethyl)silyl]-1H-pyrrole

Hazard: Irritant

Formula: C₁₃H₂₄FNSi
MDL Number: MFCD20926185
CAS Number: 156304-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR