5,6-Dihydro-imidazo[4,5,l-j-k]-[1]benzazepine-2,7-[1H,4H]-dione ≥95%
Supplier: Matrix Scientific
Synonyms:
8,9-Dihydro-2,9a-diazabenzo[cd]azulene-1,6(2H,7H)-dione
115710-1G
115710-5G
115710-10G
102897-250EA
604.62
USD
102897-250
102897-252
102897-254
5,6-Dihydro-imidazo[4,5,l-j-k]-[1]benzazepine-2,7-[1H,4H]-dione ≥95%
5,6-Dihydro-imidazo[4,5,l-j-k]-[1]benzazepine-2,7-[1H,4H]-dione
Hazard: Irritant
Formula:
C₁₁H₁₀N₂O₂ MW: 202.21 g/mol |
MDL Number:
MFCD18252621 CAS Number: 92260-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...