Diethyl-1-boc-azetidine-3-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-tert-Butyl 3,3-diethyl azetidine-1,3,3-tricarboxylate

115709-1G 115709-5G 115709-10G
102897-244EA 743.08 USD
102897-244 102897-246 102897-248
Diethyl-1-boc-azetidine-3-dicarboxylate ≥95%
Diethyl-1-boc-azetidine-3-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₂₃NO₆
MW: 301.34 g/mol
MDL Number: MFCD20126212
CAS Number: 1011479-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR