4-Bromo-2-(piperazin-1-yl)thiazole ≥97%

Supplier: Matrix Scientific

115695-1G 115695-5G 115695-10G
102899-578EA 743.08 USD
102899-578 102899-580 102899-582
4-Bromo-2-(piperazin-1-yl)thiazole ≥97%
4-Bromo-2-(piperazin-1-yl)thiazole

Hazard: Irritant

Formula: C₇H₁₀BrN₃S
MDL Number: MFCD09607694
CAS Number: 668484-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR