4-Amino-1-(3-methoxypropyl)piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3-Methoxypropyl)piperidin-4-amine

115680-1G 115680-5G 115680-10G
102899-556EA 880 USD
102899-556 102899-558 102899-560
4-Amino-1-(3-methoxypropyl)piperidine ≥95%
4-Amino-1-(3-methoxypropyl)piperidine

Hazard: Irritant

Formula: C₉H₂₀N₂O
MW: 172.27 g/mol
MDL Number: MFCD11104531
CAS Number: 179474-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR