4-Amino-1-ethylpyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

115678-1G 115678-5G
102897-200EA 1292.31 USD
102897-200 102897-202
4-Amino-1-ethylpyridin-2(1H)-one ≥95%
4-Amino-1-ethylpyridin-2(1H)-one

Hazard: Irritant

Formula: C₇H₁₀N₂O
MDL Number: MFCD19300848
CAS Number: 1310249-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR