2-Bromo-5-nitropyridin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-bromo-5-nitropyridine

115675-1G 115675-5G 115675-10G
102897-188EA 1567.69 USD
102897-188 102897-190 102897-192
2-Bromo-5-nitropyridin-4-amine ≥95%
2-Bromo-5-nitropyridin-4-amine

Hazard: Irritant

Formula: C₅H₄BrN₃O₂
MW: 218.01 g/mol
MDL Number: MFCD11100588
CAS Number: 84487-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR