1,1-Dibromo-2-chloro-2-(diethoxymethyl)cyclopropane ≥95%

Supplier: Matrix Scientific

115671-500MG 115671-1G 115671-5G
102897-166EA 495.38 USD
102897-166 102897-168 102897-170
1,1-Dibromo-2-chloro-2-(diethoxymethyl)cyclopropane ≥95%
1,1-Dibromo-2-chloro-2-(diethoxymethyl)cyclopropane

Hazard: Irritant

Formula: C₈H₁₃Br₂ClO₂
MDL Number: MFCD26406708
CAS Number: 851900-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR