Dibromoacetaldehyde diethyl acetal ≥90%

Supplier: Matrix Scientific

115670-1G 115670-5G 115670-25G
102897-160EA 330.77 USD
102897-160 102897-162 102897-164
Dibromoacetaldehyde diethyl acetal ≥90%
Dibromoacetaldehyde diethyl acetal

Hazard: Irritant

Formula: C₆H₁₂Br₂O₂
MDL Number: MFCD08457885
CAS Number: 761-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR