(S)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate ≥95%

Supplier: Matrix Scientific

115666-250MG 115666-1G
102897-142EA 330.77 USD
102897-142 102897-144
(S)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate ≥95%
(S)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD09955395
CAS Number: 127199-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR