(R)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate ≥95%

Supplier: Matrix Scientific

115665-250MG 115665-1G
102897-138EA 3295.38 USD
102897-138 102897-140
(R)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate ≥95%
(R)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD11519122
CAS Number: 127199-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR