1-Boc-3-aminomethyl-3-hydroxypiperidine ≥95%

Supplier: Matrix Scientific

115661-1G 115661-5G
102899-546EA 880 USD
102899-546 102899-548
1-Boc-3-aminomethyl-3-hydroxypiperidine ≥95%
1-Boc-3-aminomethyl-3-hydroxypiperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₃
MDL Number: MFCD18917047
CAS Number: 1308384-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR