Potassium bis(fluorosulfonyl)imide ≥98%
Supplier: Matrix Scientific
Synonyms:
Potassium bis(fluorosulphonyl)amide
115655-1G
115655-5G
115655-25G
102897-120EA
70.77
USD
102897-120
102897-122
102897-124
Potassium bis(fluorosulfonyl)imide ≥98%
Potassium bis(fluorosulfonyl)imide
Hazard: Irritant
Formula:
F₂KNO₄S₂ MW: 219.23 g/mol |
MDL Number:
MFCD17018950 CAS Number: 14984-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...