((S)-Piperidin-2-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-Piperidin-2-ylmethanol

115654-1G 115654-5G
102897-116EA 361.54 USD
102897-116 102897-118
((S)-Piperidin-2-yl)methanol ≥95%
((S)-Piperidin-2-yl)methanol

Hazard: Irritant

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD11036291
CAS Number: 41373-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR