((R)-Piperidin-2-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-Piperidin-2-ylmethanol

115653-1G 115653-5G
102897-110EA 1530.77 USD
102897-110 102897-112
((R)-Piperidin-2-yl)methanol ≥95%
((R)-Piperidin-2-yl)methanol

Hazard: Irritant

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD11036292
CAS Number: 3197-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR