6-Oxa-2-azaspiro[3.4]octane oxalate ≥97%

Supplier: Matrix Scientific

115651-250MG 115651-500MG 115651-1G
102897-096EA 893.85 USD
102897-096 102897-098 102897-100
6-Oxa-2-azaspiro[3.4]octane oxalate ≥97%
6-Oxa-2-azaspiro[3.4]octane oxalate

Hazard: Irritant

Formula: C₈H₁₃NO₅
MW: 203.2 g/mol
MDL Number: MFCD22378846
CAS Number: 1392804-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR