1-tert-Butyl-3-methyl-4,4-difluoropiperidine-1,3-dicarboxylate ≥95%

Supplier: Matrix Scientific

115645-100MG 115645-250MG 115645-500MG
102897-050EA 2527.69 USD
102897-050 102897-052 102897-054
1-tert-Butyl-3-methyl-4,4-difluoropiperidine-1,3-dicarboxylate ≥95%
1-tert-Butyl-3-methyl-4,4-difluoropiperidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₂H₁₉F₂NO₄
MDL Number: MFCD18909838
CAS Number: 1303974-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR