tert-Butyl-4-((4-fluorophenyl)(hydroxy)methyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115636-5G 115636-1G
102897-024EA 3309.23 USD
102897-024 102899-530
tert-Butyl-4-((4-fluorophenyl)(hydroxy)methyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-((4-fluorophenyl)(hydroxy)methyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₄FNO₃
MW: 309.38 g/mol
MDL Number: MFCD11041269
CAS Number: 160296-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR