tert-Butyl-4-fluoro-3-oxopiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-fluoro-3-oxopiperidine-1-carboxylate

115635-100MG 115635-500MG 115635-1G
102899-524EA 769.23 USD
102899-524 102899-526 102899-528
tert-Butyl-4-fluoro-3-oxopiperidine-1-carboxylate ≥95%
tert-Butyl-4-fluoro-3-oxopiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₆FNO₃
MW: 217.24 g/mol
MDL Number: MFCD20261388
CAS Number: 1334413-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR