tert-Butyl-3-fluoro-4-(hydroxymethyl)-4-methylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-fluoro-4-(hydroxymethyl)-4-methylpiperidine-1-carboxylate

115634-250MG 115634-1G
102899-520EA 1867.69 USD
102899-520 102899-522
tert-Butyl-3-fluoro-4-(hydroxymethyl)-4-methylpiperidine-1-carboxylate ≥95%
tert-Butyl-3-fluoro-4-(hydroxymethyl)-4-methylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂FNO₃
MW: 247.31 g/mol
MDL Number: MFCD19686589
CAS Number: 1334412-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR