(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Bromoethyl)diphenylsulphonium Triflate

115621-1G 115621-5G
102897-080EA 110.77 USD
102897-080 102897-082
(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate ≥95%
(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate

Hazard: Irritant

Formula: C₁₅H₁₄BrF₃O₃S₂
MW: 443.29 g/mol
MDL Number: MFCD12546049
CAS Number: 247129-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR