tert-Butyl-4-((benzyloxycarbonylamino)methyl)-3-fluoropiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115618-250MG 115618-500MG
102897-068EA 4393.85 USD
102897-068 102897-070
tert-Butyl-4-((benzyloxycarbonylamino)methyl)-3-fluoropiperidine-1-carboxylate ≥95%
tert-Butyl-4-((benzyloxycarbonylamino)methyl)-3-fluoropiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₉H₂₇FN₂O₄
MW: 366.44 g/mol
MDL Number: MFCD22421678
CAS Number: 1400764-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR