tert-Butyl-4-(aminomethyl)-3-fluoropiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(Aminomethyl)-3-fluoropiperidine-1-carboxylate

115614-100MG 115614-500MG
102896-980EA 1923.08 USD
102896-980 102896-982
tert-Butyl-4-(aminomethyl)-3-fluoropiperidine-1-carboxylate ≥95%
tert-Butyl-4-(aminomethyl)-3-fluoropiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₁FN₂O₂
MW: 232.3 g/mol
MDL Number: MFCD18909804
CAS Number: 1303972-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR