tert-Butyl-4-(acetamidomethyl)-3-fluoro-4-methylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115609-100MG 115609-500MG
102896-972EA 1647.69 USD
102896-972 102896-974
tert-Butyl-4-(acetamidomethyl)-3-fluoro-4-methylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-(acetamidomethyl)-3-fluoro-4-methylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₅FN₂O₃
MDL Number: MFCD22421656
CAS Number: 1400764-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR