2-Ethynyl-5-methylpyridin-3-amine ≥95%

Supplier: Matrix Scientific

115608-500MG 115608-1G 115608-5G
102896-966EA 413.85 USD
102896-966 102896-968 102896-970
2-Ethynyl-5-methylpyridin-3-amine ≥95%
2-Ethynyl-5-methylpyridin-3-amine

Hazard: Irritant

Formula: C₈H₈N₂
MDL Number: MFCD22189990
CAS Number: 1394319-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR