(S)-Pyrrolidin-3-ylmethanol ≥95%

Supplier: Matrix Scientific

115606-1G 115606-5G
102897-004EA 909.23 USD
102897-004 102897-006
(S)-Pyrrolidin-3-ylmethanol ≥95%
(S)-Pyrrolidin-3-ylmethanol

Hazard: Irritant

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD09260722
CAS Number: 110013-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR