8-Oxa-5-azaspiro[3.5]nonane ≥95%

Supplier: Matrix Scientific

115594-500MG 115594-1G
102896-952EA 3295.38 USD
102896-952 102896-954
8-Oxa-5-azaspiro[3.5]nonane ≥95%
8-Oxa-5-azaspiro[3.5]nonane

Hazard: Irritant

Formula: C₇H₁₃NO
MDL Number: MFCD17170792
CAS Number: 602326-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR