1-Oxa-7-azaspiro[3.5]nonane hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Oxa-7-azaspiro[3.5]nonane

115593-500MG 115593-1G 115593-5G
102896-946EA 1153.85 USD
102896-946 102896-948 102896-950
1-Oxa-7-azaspiro[3.5]nonane hydrochloride ≥95%
1-Oxa-7-azaspiro[3.5]nonane hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO
MW: 163.65 g/mol
MDL Number: MFCD28139317
CAS Number: 38674-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR