6-Oxa-1-azaspiro[3.3]heptane oxalate ≥95%
Supplier: Matrix Scientific
115592-500MG
115592-1G
102896-942EA
4064.62
USD
102896-942
102896-944
6-Oxa-1-azaspiro[3.3]heptane oxalate ≥95%
6-Oxa-1-azaspiro[3.3]heptane oxalate
Hazard: Irritant
Formula:
C₇H₁₁NO₅ |
MDL Number:
MFCD21364487 CAS Number: 1359655-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...