Methyl-3-(pyrrolidine-3-yl)propanoate hydrochloride ≥95%

Supplier: Matrix Scientific

115590-250MG 115590-500MG
102896-932EA 1318.46 USD
102896-932 102896-934
Methyl-3-(pyrrolidine-3-yl)propanoate hydrochloride ≥95%
Methyl-3-(pyrrolidine-3-yl)propanoate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD11111229
CAS Number: 1211593-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR