Methyl-3-(boc-amino)-3-oxetanecarboxylate ≥95%

Supplier: Matrix Scientific

115589-500MG 115589-1G 115589-5G
102896-926EA 1703.08 USD
102896-926 102896-928 102896-930
Methyl-3-(boc-amino)-3-oxetanecarboxylate ≥95%
Methyl-3-(boc-amino)-3-oxetanecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MW: 231.25 g/mol
MDL Number: MFCD22415207
CAS Number: 1363381-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR