Imidazo[1,2-a]pyridine-8-carbaldehyde ≥95%

Supplier: Matrix Scientific

115587-1G 115587-5G
102896-916EA 1292.31 USD
102896-916 102896-918
Imidazo[1,2-a]pyridine-8-carbaldehyde ≥95%
Imidazo[1,2-a]pyridine-8-carbaldehyde

Hazard: Irritant

Formula: C₈H₆N₂O
MW: 146.1500 g/mol
MDL Number: MFCD06739248
CAS Number: 136117-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR