4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine oxalate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Piperidin-4-yl)-2,2,2-trifluoroethan-1-ol ethane-1,2-dioate

115586-500MG 115586-1G
102896-912EA 1538.46 USD
102896-912 102896-914
4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine oxalate ≥95%
4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine oxalate

Hazard: Irritant

Formula: C₉H₁₄F₃NO₅
MW: 273.21 g/mol
MDL Number: MFCD18909670
CAS Number: 1298094-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR