[(3S,4R)-Rel-3-fluoro-4-piperidyl]methanol hydrochloride ≥95%

Supplier: Matrix Scientific

115584-250MG 115584-500MG 115584-1G
102896-900EA 880 USD
102896-900 102896-902 102896-904
[(3S,4R)-Rel-3-fluoro-4-piperidyl]methanol hydrochloride ≥95%
[(3S,4R)-Rel-3-fluoro-4-piperidyl]methanol hydrochloride

Hazard: Irritant

Formula: C₆H₁₂FNO
MDL Number: MFCD27937749
CAS Number: 895577-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR