3-Ethynyl-1-tosylazetidin-3-ol ≥95%

Supplier: Matrix Scientific

115583-500MG 115583-1G
102896-896EA 1538.46 USD
102896-896 102896-898
3-Ethynyl-1-tosylazetidin-3-ol ≥95%
3-Ethynyl-1-tosylazetidin-3-ol

Hazard: Irritant

Formula: C₁₂H₁₃NO₃S
MDL Number: MFCD22421707
CAS Number: 1349199-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR