2-(Boc-amino)-6-azaspiro[3.4]octane ≥95%

Supplier: Matrix Scientific

115577-500MG 115577-1G
102896-870EA 3515.38 USD
102896-870 102896-872
2-(Boc-amino)-6-azaspiro[3.4]octane ≥95%
2-(Boc-amino)-6-azaspiro[3.4]octane

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD20922774
CAS Number: 1341038-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR