3-(Tributylstannyl)-2-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

115571-1G 115571-5G 115571-25G
102899-450EA 330.77 USD
102899-450 102899-452 102899-454
3-(Tributylstannyl)-2-pyridinecarbonitrile ≥95%
3-(Tributylstannyl)-2-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₈H₃₀N₂Sn
MDL Number: MFCD22421757
CAS Number: 1416437-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR