Toluene-4-sulfonic acid tetrahydrofuran-3-ylmethyl ester ≥95%

Supplier: Matrix Scientific

115570-1G 115570-5G 115570-25G
102896-864EA 166.15 USD
102896-864 102896-866 102899-448
Toluene-4-sulfonic acid tetrahydrofuran-3-ylmethyl ester ≥95%
Toluene-4-sulfonic acid tetrahydrofuran-3-ylmethyl ester

Hazard: Irritant

Formula: C₁₂H₁₆O₄S
MDL Number: MFCD20483437
CAS Number: 15833-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR