[3-Chloro-2-(1-ethoxyethoxy)propyl] acetate ≥94%

Supplier: Matrix Scientific

115569-1G 115569-5G 115569-25G
102896-858EA 70.77 USD
102896-858 102896-860 102896-862
[3-Chloro-2-(1-ethoxyethoxy)propyl] acetate ≥94%
[3-Chloro-2-(1-ethoxyethoxy)propyl] acetate

Hazard: Irritant

Formula: C₉H₁₇ClO₄
MW: 224.69 g/mol
MDL Number: MFCD22421689
CAS Number: 85328-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR