4-Aminomethyl-2-piperidone 95

Supplier: Matrix Scientific

115568-500MG 115568-1G
102896-852EA 1813.85 USD
102896-852 102896-854
4-Aminomethyl-2-piperidone 95
4-Aminomethyl-2-piperidone

Hazard: Irritant

Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
MDL Number: MFCD17015879
CAS Number: 1234615-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR