3-(1-Naphthyl)-1-bromobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3-Bromophenyl)naphthalene

115563-1G 115563-5G 115563-25G
102896-668EA 344.62 USD
102896-668 102896-670 102896-672
3-(1-Naphthyl)-1-bromobenzene ≥95%
3-(1-Naphthyl)-1-bromobenzene

Hazard: Irritant

Formula: C₁₆H₁₁Br
MW: 283.18 g/mol
MDL Number: MFCD18415512
CAS Number: 853945-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR