4-Chloro-3-(tributylstannyl)pyridine ≥97%

Supplier: Matrix Scientific

115560-1G 115560-5G 115560-25G
102896-704EA 413.85 USD
102896-704 102896-706 102896-708
4-Chloro-3-(tributylstannyl)pyridine ≥97%
4-Chloro-3-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀ClNSn
MDL Number: MFCD20483587
CAS Number: 206115-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR